Läs mer
2,5-Bis(4-hydroxyphenyl)piperazine Dihydrochloride, TRC
Produktkod. 30335985
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30335985 500 mg
1 options
Produktkod. 30335985 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB447350500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2,5-Bis(4-hydroxyphenyl)piperazine Dihydrochloride
CAS 94572-68-6
Molekylformel C16H20Cl2N2O2
Kvantitet 500 mg
Synonym 4,4'-(2,5-Piperazinediyl)diphenol Dihydrochloride
InChI formel InChI=1S/C16H18N2O2.2ClH/c19-13-5-1-11(2-6-13)15-9-18-16(10-17-15)12-3-7-14(20)8-4-12;;/h1-8,15-20H,9-10H2;2*1H
LEDER OC(C=C1)=CC=C1C2CNC(C3=CC=C(O)C=C3)CN2
IUPAC-namn 4,4'-(piperazine-2,5-diyl)diphenol Dihydrogen chloride
Molekylvikt (g/mol) 343.25
Formel vikt 342.09