Läs mer
2,5-Bis(aminomethyl)tetrahydrofuran, TRC
Product Code. 30340363
Change view
Click to view available options
Kvantitet:
50 mg
Product Code. Kvantitet
30340363 50 mg
1 options
Product Code. 30340363 Supplier Toronto Research Chemicals Supplier No. TRCB40430050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Kemiskt namn eller material 2,5-Bis(aminomethyl)tetrahydrofuran
CAS 66918-21-6
Molekylformel C6 H14 N2 O
Kvantitet 50 mg
InChI formel InChI=1S/C6H14N2O/c7-3-5-1-2-6(4-8)9-5/h5-6H,1-4,7-8H2
LEDER NCC1CCC(CN)O1
IUPAC-namn [5-(aminomethyl)oxolan-2-yl]methanamine
Molekylvikt (g/mol) 130.19
Formel vikt 130.1106