Läs mer
2-(((5-Chloro-2-(methylamino)phenyl)(phenyl)methylene)amino)ethanol, TRC
Produktkod. 30351973
Change view
Kvantitet:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30351973 250 mg
30352244 100 mg
30368600 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30351973 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC387350250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2271-06-9
Molekylformel C16H17ClN2O
Molekylvikt (g/mol) 288.77
Synonym MAB, 2-Methylamino-5-chlor-alpha-phenylbenzylidenaminoethanol
IUPAC-namn 2-[(Z)-[[5-chloro-2-(methylamino)phenyl]-phenylmethylidene]amino]ethanol
LEDER CNc1ccc(Cl)cc1\C(=N/CCO)\c2ccccc2
Specifikationer
Kemiskt namn eller material 2-(((5-Chloro-2-(methylamino)phenyl)(phenyl)methylene)amino)ethanol
Kvantitet 250 mg
InChI formel InChI=1S/C16H17ClN2O/c1-18-15-8-7-13(17)11-14(15)16(19-9-10-20)12-5-3-2-4-6-12/h2-8,11,18,20H,9-10H2,1H3/b19-16-
Formel vikt 288.1029