Läs mer
2,5-Diacetoxy-2,5-dihydrofuran (Mixture of Isomers) (>90%), TRC
Produktkod. 30358763
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30358763 1 g
1 options
Produktkod. 30358763 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD3075751G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2,5-Diacetoxy-2,5-dihydrofuran (Mixture of Isomers)
CAS 7093-88-1
Molekylformel C8H10O5
Kvantitet 1 g
Synonym 2,5-Dihydro-2,5-furandiol 2,5-Diacetate, 2,5-Dihydro-2,5-furandiol Diacetate, 2,5-Diacetoxy-2,5-dihydrofuran, 2,5-Dihydro-2,5-diacetoxyfuran, NSC 139998
InChI formel InChI=1S/C8H10O5/c1-5(9)11-7-3-4-8(13-7)12-6(2)10/h3-4,7-8H,1-2H3
LEDER CC(=O)OC1OC(OC(=O)C)C=C1
IUPAC-namn (5-acetyloxy-2,5-dihydrofuran-2-yl) acetate
Molekylvikt (g/mol) 186.16
Formel vikt 186.0528