missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromo-3-n-dodecylthiophene, 97%, Thermo Scientific™

Catalog Number p-7037260
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
148256-63-7
C16H26Br2S
410.252
MFCD00274315
ZIZGNWAMVWNAJT-UHFFFAOYSA-N
5072754
2,5-dibromo-3-dodecylthiophene
CCCCCCCCCCCCC1=C(SC(=C1)Br)Br
This item is not returnable. View return policy
148256-63-7
C16H26Br2S
410.252
MFCD00274315
ZIZGNWAMVWNAJT-UHFFFAOYSA-N
5072754
2,5-dibromo-3-dodecylthiophene
CCCCCCCCCCCCC1=C(SC(=C1)Br)Br

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2,5-Dibromo-3-n-dodecylthiophene is used as an OLED intermediate, flavor and fragrance intermediate.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Density 1.321
Boiling Point 135°C (2.5 mmHg)
Flash Point >113°C (235°F)
Refractive Index 1.53
Quantity 250 mg
Synonym thiophene, 2,5-dibromo-3-dodecyl, 2,5-dibromo-3-laurylthiophene, 2,5-dibromo-3-dodecyl-thiophene, 2,5-bromo-3-dodecylthiophene, acmc-209cip, ksc496a6t, bidd:gt0375, thiophene,2,5-dibromo-3-dodecyl, 2,5-bis bromanyl-3-dodecyl-thiophene, 2,5-dibromo-3-dodecylthiophene
Solubility Information Slightly soluble in water.
Formula Weight 410.25
Percent Purity 97%
Chemical Name or Material 2,5-Dibromo-3-n-dodecylthiophene
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'