missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Dibromobenzaldehyde, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
2,5-Dibromobenzaldehyde is used as intermediate, it is used to produce other chemicals, it can react with Ethane-1,2-diol to get 2-(2,5-Dibromo-phenyl)-[1,3]dioxolane. The reaction occurs with reagent p-toluenesulfonic acid, solvent toluene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,5-Dibromobenzaldehyde is used as intermediate, it is used to produce other chemicals, it can react with Ethane-1,2-diol to get 2-(2,5-Dibromo-phenyl)-[1,3]dioxolane. The reaction occurs with reagent p-toluenesulfonic acid, solvent toluene.
Solubility
Soluble in water.
Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers
2,5-Dibromobenzaldehyde is used as intermediate, it is used to produce other chemicals, it can react with Ethane-1,2-diol to get 2-(2,5-Dibromo-phenyl)-[1,3]dioxolane. The reaction occurs with reagent p-toluenesulfonic acid, solvent toluene.
Solubility
Soluble in water.
Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers
Specifications
Melting Point | 88°C to 102°C |
Quantity | 1 g |
Sensitivity | Air Sensitive |
Solubility Information | Soluble in water. |
Formula Weight | 263.93 |
Percent Purity | 97% |
Chemical Name or Material | 2,5-Dibromobenzaldehyde |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'