missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichloro-p-xylene, 98%, Thermo Scientific Chemicals

Catalog Number p-7025705
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
1124-05-6
C8H8Cl2
175.052
MFCD00000610
UTGSRNVBAFCOEU-UHFFFAOYSA-N
2,5-dichloro-p-xylene, 2,5-dichloro-1,4-dimethylbenzene, benzene, 1,4-dichloro-2,5-dimethyl, p-xylene, 2,5-dichloro, unii-7p8m86zob2, 2,5-dichloro-para-xylene, p-xylene,5-dichloro, p-xylene,2,5-dichloro, acmc-2099eq, dsstox_cid_24780
70755
1,4-dichloro-2,5-dimethylbenzene
CC1=CC(=C(C=C1Cl)C)Cl
This item is not returnable. View return policy
1124-05-6
C8H8Cl2
175.052
MFCD00000610
UTGSRNVBAFCOEU-UHFFFAOYSA-N
2,5-dichloro-p-xylene, 2,5-dichloro-1,4-dimethylbenzene, benzene, 1,4-dichloro-2,5-dimethyl, p-xylene, 2,5-dichloro, unii-7p8m86zob2, 2,5-dichloro-para-xylene, p-xylene,5-dichloro, p-xylene,2,5-dichloro, acmc-2099eq, dsstox_cid_24780
70755
1,4-dichloro-2,5-dimethylbenzene
CC1=CC(=C(C=C1Cl)C)Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 68°C to 70°C
Boiling Point 221°C to 223°C
Quantity 25 g
Formula Weight 175.06
Percent Purity 98%
Chemical Name or Material 2,5-Dichloro-p-xylene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'