Läs mer
2,5-Dihydroxybenzoic Acid Butylamine Salt [Matrix for MALDI-TOF/MS], TRC
Produktkod. 30369403
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30369403 10 mg
1 options
Produktkod. 30369403 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD67919510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2,5-Dihydroxybenzoic Acid Butylamine Salt [Matrix for MALDI-TOF/MS]
CAS 666174-80-7
Molekylformel C7H6O4 . C4H11N
Kvantitet 10 mg
Synonym Benzoic acid, 2,5-dihydroxy-, compd. with 1-butanamine (1:1) (9CI, ACI), 2,5-Dihydroxybenzoic acid-butylamine (1:1) salt, Butylammonium 2,5-dihydroxybenzoate, 2,5-Dihydroxybenzoic acid butylamine salt, 2,5-Dihydroxybenzoic acid 1-butylamine salt
InChI formel InChI=1S/C7H6O4.C4H11N/c8-4-1-2-6(9)5(3-4)7(10)11;1-2-3-4-5/h1-3,8-9H,(H,10,11);2-5H2,1H3
LEDER CCCCN.OC(=O)c1cc(O)ccc1O
IUPAC-namn butan-1-amine;2,5-dihydroxybenzoic acid
Molekylvikt (g/mol) 227.26
Formel vikt 227.1158