Läs mer
2,5-Dimethyl-3-(2-methylphenyl)-1-propylimidazolini-4-one Hydrochloride, TRC
Produktkod. 30373973
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30373973 50 mg
1 options
Produktkod. 30373973 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD47705650MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material rac-trans-2,5-Dimethyl-3-(2-methylphenyl)-1-propylimidazolini-4-one Hydrochloride
CAS 677029-53-7
Molekylformel C15 H22 N2 O . H Cl
Kvantitet 50 mg
Synonym 4-Imidazolidinone, 2,5-dimethyl-3-(2-methylphenyl)-1-propyl-, hydrochloride (1:1) (ACI), 4-Imidazolidinone, 2,5-dimethyl-3-(2-methylphenyl)-1-propyl-, hydrochloride (1:1), 4-Imidazolidinone, 2,5-dimethyl-3-(2-methylphenyl)-1-propyl-, monohydrochloride (9CI), 2,5-Dimethyl-3-(2-methylphenyl)-1-propylimidazolidin-4-one hydrochloride
InChI formel InChI=1S/C15H22N2O.ClH/c1-5-10-16-12(3)15(18)17(13(16)4)14-9-7-6-8-11(14)2;/h6-9,12-13H,5,10H2,1-4H3;1H
LEDER CC1=C(N2C(C)N(CCC)C(C)C2=O)C=CC=C1.Cl
IUPAC-namn 2,5-dimethyl-3-(2-methylphenyl)-1-propylimidazolidin-4-one;hydrochloride
Molekylvikt (g/mol) 282.8089
Formel vikt 282.1499