Läs mer
2,5-Dimethyl Celecoxib, TRC
Produktkod. 30374066
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30374066 25 mg
30383219 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30374066 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD47089025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 457639-26-8
Molekylformel C18 H16 F3 N3 O2 S
Molekylvikt (g/mol) 395.3987
Synonym 2,5-Dimethylcelecoxib, Benzenesulfonamide, 4-[5-(2,5-dimethylphenyl)-3-(trifluoromethyl)-1H-pyrazol-1-yl]-, 4-[5-(2,5-Dimethylphenyl)-3-(trifluoromethyl)-1H-pyrazol-1-yl]benzenesulfonamide
IUPAC-namn 4-[5-(2,5-dimethylphenyl)-3-(trifluoromethyl)pyrazol-1-yl]benzenesulfonamide
LEDER Cc1ccc(C)c(c1)c2cc(nn2c3ccc(cc3)S(=O)(=O)N)C(F)(F)F
Specifikationer
Kemiskt namn eller material 2,5-Dimethylcelecoxib
Kvantitet 25 mg
InChI formel InChI=1S/C18H16F3N3O2S/c1-11-3-4-12(2)15(9-11)16-10-17(18(19,20)21)23-24(16)13-5-7-14(8-6-13)27(22,25)26/h3-10H,1-2H3,(H2,22,25,26)
Formel vikt 395.0915