Learn More
2',5'-Dimethylacetophenone, 97%
Product Code. 11470283
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11470283 100 g 100g
11450283 5 g 5g
11460283 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11470283 Supplier Thermo Scientific Alfa Aesar Supplier No. A11432.22
 Restricted Item
Beräknad leverans: 02-12-2026
Add to basket
CAS: 2142-73-6 | C10H12O | 148.205 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2142-73-6
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.205
MDL Number MFCD00014988
InChI Key AWKBVLVKQQRRFQ-UHFFFAOYSA-N
Synonym 2',5'-dimethylacetophenone, 2,5-dimethylacetophenone, 1-2,5-dimethylphenyl ethanone, 1-2,5-dimethylphenyl ethan-1-one, ethanone, 1-2,5-dimethylphenyl, acetophenone, 2',5'-dimethyl, 1-acetyl-2,5-dimethylbenzene, 2-acetyl-p-xylene, acmc-1cliy, 2',5'dimethylacetophenone
PubChem CID 75061
IUPAC Name 1-(2,5-dimethylphenyl)ethanone
SMILES CC1=CC(=C(C=C1)C)C(=O)C
Specifications
Boiling Point 230°C to 231°C
Flash Point 95°C (203°F)
Odor Characteristic
Quantity 100 g
Beilstein 2042139
Formula Weight 148.21
Percent Purity ≥98%
Chemical Name or Material 2',5'-Dimethylacetophenone