Learn More
2,5-Dimethylphenol, 97%, Thermo Scientific™
Catalog Number p-7023822
Click to view available options
Quantity:
250 g
1000 g
95-87-4
C8H10O
122.17
MFCD00002237
NKTOLZVEWDHZMU-UHFFFAOYSA-N
2,5-xylenol, p-xylenol, phenol, 2,5-dimethyl, 3,6-dimethylphenol, 6-methyl-m-cresol, 3,6-xylenol, 2,5-dimethyl phenol, 2,5-dmp, 1,2,5-xylenol, 1-hydroxy-2,5-dimethylbenzene
7267
2,5-dimethylphenol
CC1=CC=C(C)C(O)=C1
95-87-4
C8H10O
122.17
MFCD00002237
NKTOLZVEWDHZMU-UHFFFAOYSA-N
2,5-xylenol, p-xylenol, phenol, 2,5-dimethyl, 3,6-dimethylphenol, 6-methyl-m-cresol, 3,6-xylenol, 2,5-dimethyl phenol, 2,5-dmp, 1,2,5-xylenol, 1-hydroxy-2,5-dimethylbenzene
7267
2,5-dimethylphenol
CC1=CC=C(C)C(O)=C1
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Used to make phenolic resins and (in isomeric mixtures) as an antioxidant for lubricating oils, gasoline, and elastomers. Also used as perfuming agents.
Solubility
Soluble in water.
Notes
Store in a tightly closed container in a cool, dry, well-ventilated area. Store away from strong oxidizing agents. Store under dry inert gas.
TRUSTED_SUSTAINABILITY
Melting Point 74°C to 78°C
UN Number UN2261
Beilstein 1099260
Merck Index 14,10082
Solubility Information Soluble in water.
Formula Weight 122.17
Percent Purity 97%
Density 0.971
Boiling Point 211°C to 213°C
Quantity 250 g
Chemical Name or Material 2,5-Dimethylphenol