Learn More
2,5-Dimethylphenylboronic acid, 97%, Thermo Scientific™
Catalog Number p-4228624
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
85199-06-0
C8H11BO2
149.98
MFCD01863525
OOMZKLJLVGQZGV-UHFFFAOYSA-N
2,5-dimethylphenyl boronic acid, 2,5-dimethylbenzeneboronic acid, 2,5-dimethylphenylboronicacid, 2,5-dimethylphenyl boranediol, p-xylene-2-boronic acid, boronic acid, 2,5-dimethylphenyl, pubchem1831, p-xyleneboronic acid, ksc448a1r, 2,5-dimehylphenylboronic acid
2734347
(2,5-dimethylphenyl)boronic acid
CC1=CC=C(C)C(=C1)B(O)O
85199-06-0
C8H11BO2
149.98
MFCD01863525
OOMZKLJLVGQZGV-UHFFFAOYSA-N
2,5-dimethylphenyl boronic acid, 2,5-dimethylbenzeneboronic acid, 2,5-dimethylphenylboronicacid, 2,5-dimethylphenyl boranediol, p-xylene-2-boronic acid, boronic acid, 2,5-dimethylphenyl, pubchem1831, p-xyleneboronic acid, ksc448a1r, 2,5-dimehylphenylboronic acid
2734347
(2,5-dimethylphenyl)boronic acid
CC1=CC=C(C)C(=C1)B(O)O
Melting Point 186.0°C to 191.0°C
CAS Min % 96.0
CAS Max % 100.0
Quantity 1 g
Packaging Glass bottle
Assay Percent Range 96% min. (HPLC)
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol
Formula Weight 149.98
Percent Purity 97%
Chemical Name or Material 2, 5-Dimethylphenylboronic acid
compliance-icons
Product Identifier
  • 2,5-Dimethylphenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.