Läs mer
2,5-Dinitrotoluene, TRC
Produktkod. 30137194
Change view
Kvantitet:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137194 250 mg
30133640 1 g
30008771 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137194 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD426810250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 619-15-8
Molekylformel C7 H6 N2 O4
Molekylvikt (g/mol) 182.13
Synonym Toluene, 2,5-dinitro- (8CI), 2-Methyl-1,4-dinitrobenzene, 1-Methyl-2,5-dinitrobenzene, 2,5-DNT, 2,5-Dinitrotoluene, 3-Methyl-1,4-dinitrobenzene, NSC 159120, Benzene, 2-methyl-1,4-dinitro-
IUPAC-namn 2-methyl-1,4-dinitrobenzene
LEDER Cc1cc(ccc1[N+](=O)[O-])[N+](=O)[O-]
Specifikationer
Kemiskt namn eller material 2,5-Dinitrotoluene
Anteckningar om renhetsgrad HPLC
Kvantitet 250 mg
InChI formel InChI=1S/C7H6N2O4/c1-5-4-6(8(10)11)2-3-7(5)9(12)13/h2-4H,1H3
Formel vikt 182.0328
Procent renhet >95%