Learn More
2,5-Dioxopyrrolidin-1-yl 3',6'-dihydroxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-5-carboxylate, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 92557-80-7 |
| Molecular Formula | C25H15NO9 |
| MDL Number | MFCD00077322 |
| Quantity | 1 g |
| SMILES | O=C1OC2(C3=C(OC4=C2C=CC(O)=C4)C=C(O)C=C3)C5=C1C=C(C(ON6C(CCC6=O)=O)=O)C=C5 |
| Molecular Weight (g/mol) | 473.39 |
| Percent Purity | 97% |
| Chemical Name or Material | 2,5-Dioxopyrrolidin-1-yl 3',6'-dihydroxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-5-carboxylate |