Läs mer
2-(5-Mercaptotetrazole-1-yl)ethanol, TRC
Produktkod. 30474042
Change view
Kvantitet:
1 g
5 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30474042 50 g
30475113 1 g
30472262 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30474042 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM22546050G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 56610-81-2
Molekylformel C3 H6 N4 O S
Molekylvikt (g/mol) 146.17
Synonym 1,2-Dihydro-1-(2-hydroxyethyl)-5H-tetrazole-5-thione, 1-(2-Hydroxyethyl)-1H-tetrazole-5-thiol, 1-(2-Hydroxyethyl)-5-mercapto-1H-tetrazole, 1-(2-Hydroxyethyl)-5-mercaptotetrazole, 1-(2-Hydroxyethyl)tetrazole-5-thiol, 5-Mercapto-1-(2-hydroxyethyl)-1H-tetrazole, 5-Mercapto-1-tetrazoleethanol, [1-(2-Hydroxyethyl)-1H-1H-tetrazol-5-yl]thiol
IUPAC-namn 2-(5-sulfanyltetrazol-1-yl)ethanol
LEDER OCCn1nnnc1S
Specifikationer
Kemiskt namn eller material 2-(5-Mercaptotetrazole-1-yl)ethanol
Kvantitet 50 g
InChI formel InChI=1S/C3H6N4OS/c8-2-1-7-3(9)4-5-6-7/h8H,1-2H2,(H,4,6,9)
Formel vikt 146.0262