missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(5-Methyl-2-thienyl)thiazolidine-4-carboxylic acid, 97%

Brand:  Alfa Aesar™ H62089.06

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15497006

  • 11418.38 SEK / 5g


Estimated Shipment date
from Supplier 17-04-2019
Add to basket

Specifications

Chemical Identifiers

MFCD08559334

Specifications

2-(5-Methyl-2-thienyl)thiazolidine-4-carboxylic acid
MFCD08559334
Solid
5g
229.32
97%
Videos
SDS