Läs mer
2,5-Pyrazinedipropanoic Acid Dimethyl Ester, TRC
Produktkod. 30416596
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30416596 10 mg
1 options
Produktkod. 30416596 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP84083510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2,5-Pyrazinedipropanoic Acid Dimethyl Ester
CAS 77479-01-7
Molekylformel C12H16N2O4
Kvantitet 10 mg
Synonym 2,5-Pyrazinedipropanoic Acid 2,5-Dimethyl Ester, 2,5-beta-Carboxyethyl Pyrazine Dimethylester
InChI formel InChI=1S/C12H16N2O4/c1-17-11(15)5-3-9-7-14-10(8-13-9)4-6-12(16)18-2/h7-8H,3-6H2,1-2H3
LEDER COC(=O)CCc1cnc(CCC(=O)OC)cn1
IUPAC-namn methyl 3-[5-(3-methoxy-3-oxopropyl)pyrazin-2-yl]propanoate
Molekylvikt (g/mol) 252.27
Formel vikt 252.111