missing translation for 'onlineSavingsMsg'
Läs mer

Cibacron Brilliant Yellow 3G-P, Technical Grade, TRC

Product Code. 30179152
Change view
Click to view available options
Kvantitet:
1 g
2.5 g
5 g
Unit Size:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet unitSize
30179152 2.5 g 2.5g
30179410 5 g -
30179039 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179152 Supplier Toronto Research Chemicals Supplier No. TRCC5357332.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50662-99-2
Molekylformel C25H15Cl3N9Na3O10S3
Molekylvikt (g/mol) 872.96
Synonym 4-[[4-Chloro-6-[(4-sulfophenyl)amino]-1,3,5-triazin-2-yl]amino]-2-[[1-(2,5-dichloro-4-sulfophenyl)-4,5-dihydro-3-methyl-5-oxo-1H-pyrazol-4-yl]azo]-benzenesulfonic Acid trisodium salt, C.I. 18972C.I. Reactive Yellow 2Cibacron Brilliant Yellow 3G-PCibacron Yellow 3GHelaktyn Yellow D 5GNKP Cion Yellow P-VKayacion Yellow P 5GKayacion Yellow P 5G Liquid 20Kayacion Yellow P 5G Liquid 33Ostazin Brilliant Yellow H 5GOstazin Yellow H 5GProcion Brilliant Yellow H 5GProcion Golden Yellow H 5GProcion Yellow H 5GProcion Yellow P 5GNReactive Flavine Yellow K 6GReactive Yellow 2Reactive Yellow K 6GReactive Yellow M 3GPSumifix Brilliant Yellow H 3GXiron Brilliant Yellow 3G-HDYellow P 5G
IUPAC-namn trisodium;2,5-dichloro-4-[4-[[5-[[4-chloro-6-(4-sulfonatoanilino)-1,3,5-triazin-2-yl]amino]-2-sulfonatophenyl]diazenyl]-3-methyl-5-oxo-4H-pyrazol-1-yl]benzenesulfonate
LEDER [Na+].[Na+].[Na+].CC1=NN(C(=O)C1N=Nc2cc(Nc3nc(Cl)nc(Nc4ccc(cc4)S(=O)(=O)[O-])n3)ccc2S(=O)(=O)[O-])c5cc(Cl)c(cc5Cl)S(=O)(=O)[O-]

Specifications

Kemiskt namn eller material Cibacron Brilliant Yellow 3G-P, Technical Grade
Kvantitet 2.5 g
InChI formel InChI=1S/C25H18Cl3N9O10S3.3Na/c1-11-21(22(38)37(36-11)18-9-16(27)20(10-15(18)26)50(45,46)47)35-34-17-8-13(4-7-19(17)49(42,43)44)30-25-32-23(28)31-24(33-25)29-12-2-5-14(6-3-12)48(39,40)41;;;/h2-10,21H,1H3,(H,39,40,41)(H,42,43,44)(H,45,46,47)(H2,29,30,31,32,33);;;/q;3*+1/p-3
Formel vikt 870.8863
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.