Läs mer
2,6-Anhydro-5-deoxy-D-lyxo-hex-5-enonic Acid Methyl Ester 3,4-Diacetate, TRC
Produktkod. 30331210
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30331210 50 mg
1 options
Produktkod. 30331210 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA63820050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2,6-Anhydro-5-deoxy-D-lyxo-hex-5-enonic Acid Methyl Ester 3,4-Diacetate
CAS 57690-62-7
Molekylformel C11H14O7
Kvantitet 50 mg
Synonym 2,6-Anhydro-5-deoxy-D-lyxo-hex-5-enonic Acid Methyl Ester Diacetate
InChI formel InChI=1S/C11H14O7/c1-6(12)17-8-4-5-16-10(11(14)15-3)9(8)18-7(2)13/h4-5,8-10H,1-3H3/t8-,9+,10+/m1/s1
LEDER COC(=O)[C@H]1OC=C[C@@H](OC(=O)C)[C@@H]1OC(=O)C
IUPAC-namn methyl (2S,3S,4R)-3,4-diacetyloxy-3,4-dihydro-2H-pyran-2-carboxylate
Molekylvikt (g/mol) 258.22
Formel vikt 258.074