missing translation for 'onlineSavingsMsg'
Learn More

2,6-Bis(dodecyloxy)benzaldehyde, 97%, Thermo Scientific Chemicals

Catalog Number p-7055492
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
1198467-57-0
C31H54O3
474.77
MFCD25372091
QPAKGKFSBBAHMS-UHFFFAOYSA-N
2,6-bis dodecyloxy benzaldehyde, 2,6-di dodecyloxy benzaldehyde
75201262
2,6-didodecoxybenzaldehyde
CCCCCCCCCCCCOC1=C(C(=CC=C1)OCCCCCCCCCCCC)C=O
This item is not returnable. View return policy
1198467-57-0
C31H54O3
474.77
MFCD25372091
QPAKGKFSBBAHMS-UHFFFAOYSA-N
2,6-bis dodecyloxy benzaldehyde, 2,6-di dodecyloxy benzaldehyde
75201262
2,6-didodecoxybenzaldehyde
CCCCCCCCCCCCOC1=C(C(=CC=C1)OCCCCCCCCCCCC)C=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Yellow
Quantity 250 mg
Beilstein 20848961
Formula Weight 474.77
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2,6-Bis(dodecyloxy)benzaldehyde

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'