Läs mer
2,6-Di-tert-butyl-4-ethylphenol, TRC
Produktkod. 30136868
Change view
Kvantitet:
1 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136868 100 g
30010896 25 g
17960805 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136868 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD493890100G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 4130-42-1
Molekylformel C16 H26 O
Molekylvikt (g/mol) 234.38
Synonym 2,6-Di-tert-butyl-4-ethylphenol, Phenol, 2,6-di-tert-butyl-4-ethyl- (6CI,7CI,8CI), 2,6-Bis(1,1-dimethylethyl)-4-ethylpheno, 2,6-Di-tert-butyl-p-ethylphenol, 2,6-Ditert-butyl-4-ethylphenol, 4-Ethyl-2,6-di-tert-butylphenol, BHEB, DBEP, Ionol 2, NSC 14453, Noclizer M 17, Nocrac M 17, Sandant 425, Yoshinox 250
IUPAC-namn 2,6-ditert-butyl-4-ethylphenol
LEDER CCc1cc(c(O)c(c1)C(C)(C)C)C(C)(C)C
Specifikationer
Kemiskt namn eller material 2,6-Di-tert-butyl-4 ethylphenol
Kvantitet 100 g
InChI formel InChI=1S/C16H26O/c1-8-11-9-12(15(2,3)4)14(17)13(10-11)16(5,6)7/h9-10,17H,8H2,1-7H3
Formel vikt 234.1984