missing translation for 'onlineSavingsMsg'
Läs mer

2,6-Di-tert-butyl-4-nitrophenol, TRC

Produktkod. 30371161
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30371161 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30371161 Supplier Toronto Research Chemicals Supplier No. TRCD493720500MG

Please to purchase this item. Need a web account? Register with us today!

Denna vara kräver en prisuppdatering innan den beställs.

Begär ett pris för denna vara här.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 2,6-Di-tert-butyl-4-nitrophenol
CAS 728-40-5
Molekylformel C14 H21 N O3
Kvantitet 500 mg
Synonym 2,6-bis(1,1-Dimethylethyl)-4-nitrophenol, 4-Nitro-2,6-bis-(1,1-dimethylethyl)phenol, 4-Nitro-2,6-di-tert-butylphenol, BAY 28589, Bayer 28589, NSC 81682
InChI formel InChI=1S/C14H21NO3/c1-13(2,3)10-7-9(15(17)18)8-11(12(10)16)14(4,5)6/h7-8,16H,1-6H3
LEDER CC(C)(C)c1cc(cc(c1O)C(C)(C)C)[N+](=O)[O-]
IUPAC-namn 2,6-ditert-butyl-4-nitrophenol
Molekylvikt (g/mol) 251.32
Formel vikt 251.1521
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.