Läs mer
2,6-Diamino-1-methyl-4(1H)-pyrimidinone, TRC
Product Code. 30355633
Change view
Click to view available options
Kvantitet:
500 mg
Product Code. Kvantitet
30355633 500 mg
1 options
Product Code. 30355633 Supplier Toronto Research Chemicals Supplier No. TRCD416548500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Kemiskt namn eller material 2,6-Diamino-1-methyl-4(1H)-pyrimidinone
CAS 51093-34-6
Molekylformel C5H8N4O
Kvantitet 500 mg
Synonym NSC 618274
InChI formel InChI=1S/C5H8N4O/c1-9-3(6)2-4(10)8-5(9)7/h2H,6H2,1H3,(H2,7,8,10)
LEDER CN1C(=CC(=O)N=C1N)N
IUPAC-namn 2,6-diamino-1-methylpyrimidin-4-one
Molekylvikt (g/mol) 140.14
Formel vikt 140.0698