Läs mer
2,6-Diaminopurine Arabinoside, TRC
Produktkod. 30366615
Change view
Kvantitet:
500 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30366615 5 g
30352884 1 g
30351238 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30366615 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4167385G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 34079-68-0
Molekylformel C10H14N6O4
Molekylvikt (g/mol) 282.26
Synonym 9-beta-D-Arabinofuranosyl-9H-purine-2,6-diamine, 2,6-Diamino-9-beta-D-arabinofuranosylpurine, 9-beta-D-Arabinofuranosyl-2,6-diaminopurine
IUPAC-namn (2R,3S,4S,5R)-2-(2,6-diaminopurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
LEDER Nc1nc(N)c2ncn([C@@H]3O[C@H](CO)[C@@H](O)[C@@H]3O)c2n1
Specifikationer
Kemiskt namn eller material 2,6-Diaminopurine Arabinoside
Kvantitet 5 g
InChI formel InChI=1S/C10H14N6O4/c11-7-4-8(15-10(12)14-7)16(2-13-4)9-6(19)5(18)3(1-17)20-9/h2-3,5-6,9,17-19H,1H2,(H4,11,12,14,15)/t3-,5-,6+,9-/m1/s1
Formel vikt 282.1077