missing translation for 'onlineSavingsMsg'
Learn More

2,6-Diaminopyridine, 98%, Thermo Scientific Chemicals

Catalog Number p-100368
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
141-86-6
C5H7N3
109.13
MFCD00006329
VHNQIURBCCNWDN-UHFFFAOYSA-N
2,6-diaminopyridine, 2,6-pyridinediamine, 2,6-diamino pyridine, pyridine, 2,6-diamino, dap amine, pyridine-2,6-diyldiamine, unii-k89ab8ckg6, ccris 6682, k89ab8ckg6, dsstox_cid_20127
8861
pyridine-2,6-diamine
C1=CC(=NC(=C1)N)N
This item is not returnable. View return policy
141-86-6
C5H7N3
109.13
MFCD00006329
VHNQIURBCCNWDN-UHFFFAOYSA-N
2,6-diaminopyridine, 2,6-pyridinediamine, 2,6-diamino pyridine, pyridine, 2,6-diamino, dap amine, pyridine-2,6-diyldiamine, unii-k89ab8ckg6, ccris 6682, k89ab8ckg6, dsstox_cid_20127
8861
pyridine-2,6-diamine
C1=CC(=NC(=C1)N)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 115°C to 123°C
Color Beige to Brown-Gray
Boiling Point 285°C
Flash Point 155°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (C5H3N)(NH2)2
Quantity 25 g
Beilstein 22, I, 647
Solubility Information Solubility in water: 9.9g/100 mL (20°C)
Formula Weight 109.13
Percent Purity 98%
Physical Form Crystalline Powder or Flakes
Chemical Name or Material 2, 6-Diaminopyridine, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'