Learn More
2,6-Dibromo-4-methylaniline, 95%, BLD Pharm™
Product Code. 31432491
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31432491 25 g
31414430 5 g
31432187 500 g
31419219 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 31432491 Supplier BLD Pharm Supplier No. BD1048825g
 Restricted Item
Beräknad leverans: 25-11-2026
Specifications
CAS 6968-24-7
Molecular Formula C7H7Br2N
MDL Number MFCD00007641
Quantity 25 g
SMILES C1=C(C)C=C(C(=C1Br)N)Br
Molecular Weight (g/mol) 264.95
Percent Purity 95%
Chemical Name or Material 2,6-Dibromo-4-methylaniline