missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dibromoaniline, 97%, Thermo Scientific Chemicals

Catalog Number p-102903
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
608-30-0
C6H5Br2N
250.92
MFCD00007638
XIRRDAWDNHRRLB-UHFFFAOYSA-N
benzenamine, 2,6-dibromo, 2,6-dibromophenylamine, 2,6-dibromobenzenamine, aniline, 2,6-dibromo, 2,6-dibromo-phenylamine, 2,6-dibromo-aniline, acmc-1aycf, 2,6-dibromoaniline, 2,6-dibromo-phenyl-amine, ksc356c3l
69098
2,6-dibromoaniline
NC1=C(Br)C=CC=C1Br
This item is not returnable. View return policy
608-30-0
C6H5Br2N
250.92
MFCD00007638
XIRRDAWDNHRRLB-UHFFFAOYSA-N
benzenamine, 2,6-dibromo, 2,6-dibromophenylamine, 2,6-dibromobenzenamine, aniline, 2,6-dibromo, 2,6-dibromo-phenylamine, 2,6-dibromo-aniline, acmc-1aycf, 2,6-dibromoaniline, 2,6-dibromo-phenyl-amine, ksc356c3l
69098
2,6-dibromoaniline
NC1=C(Br)C=CC=C1Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 79.0°C to 84.0°C
Color Gray or White to Yellow
Boiling Point 262.0°C to 264.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula Br2C6H3NH2
Quantity 10 g
Beilstein 12, 659
Formula Weight 250.92
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2, 6-Dibromoaniline, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 2,6-Dibromoaniline
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'