missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-dichloro-1,3-benzothiazole, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
25 g
Specifications
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Formula Weight | 204.08 |
Percent Purity | 97% |
Chemical Name or Material | 2, 6-Dichloro-1, 3-benzothiazole |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'