missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloro-4-isocyanatopyridine, 90%, Thermo Scientific Chemicals

Catalog Number p-7077368
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
159178-03-7
C6H2Cl2N2O
189
OCDFIQXUCLQQHY-UHFFFAOYSA-N
2781658
2,6-dichloro-4-isocyanatopyridine
C1=C(C=C(N=C1Cl)Cl)N=C=O
This item is not returnable. View return policy
159178-03-7
C6H2Cl2N2O
189
OCDFIQXUCLQQHY-UHFFFAOYSA-N
2781658
2,6-dichloro-4-isocyanatopyridine
C1=C(C=C(N=C1Cl)Cl)N=C=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 38.0°C to 51.0°C
Infrared Spectrum Authentic
Assay Percent Range 89% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Synonym 2,6-dichloro-4-pyridyl isocyanate, 2,6-dichloropyridin-4-isocyanate, 2,6-dichloropyridin-4-yl isocyanate, pyridine,2,6-dichloro-4-isocyanato, pyridine, 2,6-dichloro-4-isocyanato, 2,6-dichloropyridin-4-ylisocyanate, pyridine, 2,6-dichloro-4-isocyanato-9ci
Formula Weight 189
Percent Purity 90%
Chemical Name or Material 2,6-Dichloro-4-isocyanatopyridine
compliance-icons compliance-icons
Product Identifier
  • 2,6-Dichloro-4-isocyanatopyridine, 90%
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'