missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,6-Dichloro-4-methylpyridine, 95%

Brand:  Alfa Aesar™ H61767.03

Additional Details : CAS Number : 39621-00-6 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15438338

  • 924.61 SEK / 1g


In stock, ready to ship: 3
Add to basket

Specifications

Chemical Identifiers

39621-00-6
162.013
GTHRJKYVJZJPCF-UHFFFAOYSA-N
10898997
CC1=CC(=NC(=C1)Cl)Cl
C6H5Cl2N
MFCD09264302
2,6-dichloro-4-picoline, 2,6-dichloro-4-methyl-pyridine, pyridine, 2,6-dichloro-4-methyl, pubchem5534, acmc-209j6i, pydc11, pyridine,2,6-dichloro-4-methyl, 2,6-bis chloranyl-4-methyl-pyridine
2,6-dichloro-4-methylpyridine

Specifications

2,6-Dichloro-4-methylpyridine
1g
C6H5Cl2N
2,6-dichloro-4-picoline, 2,6-dichloro-4-methyl-pyridine, pyridine, 2,6-dichloro-4-methyl, pubchem5534, acmc-209j6i, pydc11, pyridine,2,6-dichloro-4-methyl, 2,6-bis chloranyl-4-methyl-pyridine
CC1=CC(=NC(=C1)Cl)Cl
162.013
162.02
95%
White
39621-00-6
MFCD09264302
GTHRJKYVJZJPCF-UHFFFAOYSA-N
2,6-dichloro-4-methylpyridine
10898997
Solid
Videos
SDS Documents