missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloro-4-pyridyl isocyanate, 97%, Thermo Scientific™

Catalog Number p-4510346
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
159178-03-7
C6H2Cl2N2O
188.995
OCDFIQXUCLQQHY-UHFFFAOYSA-N
2781658
2,6-dichloro-4-isocyanatopyridine
C1=C(C=C(N=C1Cl)Cl)N=C=O
This item is not returnable. View return policy
159178-03-7
C6H2Cl2N2O
188.995
OCDFIQXUCLQQHY-UHFFFAOYSA-N
2781658
2,6-dichloro-4-isocyanatopyridine
C1=C(C=C(N=C1Cl)Cl)N=C=O
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 2,6-dichloro-4-pyridyl isocyanate, 2,6-dichloropyridin-4-isocyanate, 2,6-dichloropyridin-4-yl isocyanate, pyridine,2,6-dichloro-4-isocyanato, pyridine, 2,6-dichloro-4-isocyanato, 2,6-dichloropyridin-4-ylisocyanate, pyridine, 2,6-dichloro-4-isocyanato-9ci
Formula Weight 189
Percent Purity 90%
Chemical Name or Material 2, 6-Dichloro-4-isocyanatopyridine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'