Learn More
2,6-Dichloro-4-(trifluoromethyl)aniline, 98%, BLD Pharm™
Product Code. 30948829
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
500 g
1 kg
Unit Size:
100g
10g
1kg
25g
500g
5g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30948829 100 g 100g
30935438 25 g 25g
30947959 1 kg 1kg
30948010 500 g 500g
30933240 5 g 5g
30939132 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 30948829 Supplier BLD Pharm Supplier No. BD10261100g
 Restricted Item
Beräknad leverans: 27-11-2026
Specifications
CAS 24279-39-8
Molecular Formula C7H4Cl2F3N
MDL Number MFCD00052918
Quantity 100 g
SMILES C1=C(C=C(C(=C1Cl)N)Cl)C(F)(F)F
Molecular Weight (g/mol) 230.01
Percent Purity 98%
Chemical Name or Material 2,6-Dichloro-4-(trifluoromethyl)aniline