Learn More
Alfa Aesar™ 2,6-Difluoro-N-(3,5-difluorophenyl)benzamide, 97%
Brand:  Alfa Aesar™ H59800.03
Additional Details : Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15486208
  • 5659.56 SEK / 1g
Estimated Shipment date
from Supplier 01-02-2018
Add to basket
Specifications
Chemical Identifiers
2,6-Difluoro-N-(3,5-difluorophenyl)benzamide
269.19
MFCD11774878
Specifications
2,6-Difluoro-N-(3,5-difluorophenyl)benzamide
Solid
269.19
1g
97%
MFCD11774878