Läs mer
2,6-Dihydro-2,2,4-trimethyl-6-quinolone, TRC
Produktkod. 17972715
Change view
Kvantitet:
1 mg
2.5 mg
5 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
17972715 5 mg -
17924865 1 mg -
17931615 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17972715 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4499005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 4071-18-5
Molekylformel C12 H13 N O
Molekylvikt (g/mol) 187.24
Synonym Ethoxyquin Quinone Imine, 6(2H)-Quinolone, 2,2,4-trimethyl- (7CI,8CI), 2,2,4-Trimethyl-6(2H)-quinolinone, 2,2,4-Trimethyl-2,6-dihydro-6-quinolinone, 2,6-Dihydro-2,2,4-trimethyl-6-quinolone, 6(2H)-Quinolinone, 2,2,4-trimethyl-
IUPAC-namn 2,2,4-trimethylquinolin-6-one
LEDER CC1=CC(C)(C)N=C2C=CC(=O)C=C12
Specifikationer
Kemiskt namn eller material 2,6-Dihydro-2,2,4-trimethyl-6-quinolone
Kvantitet 5 mg
InChI formel InChI=1S/C12H13NO/c1-8-7-12(2,3)13-11-5-4-9(14)6-10(8)11/h4-7H,1-3H3
Formel vikt 187.0997