Learn More
2,6-Diiodo-4-nitrophenol, TRC
Product Code. 30372226
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity Unit Size
30372226 1 g 1g
1 options
Product Code. 30372226 Supplier Toronto Research Chemicals Supplier No. TRCD4550981G
 Restricted Item
Beräknad leverans: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2,6-Diiodo-4-nitrophenol
CAS 305-85-1
Molecular Formula C6H3I2NO3
Quantity 1 g
Synonym 4-Nitro-2,6-diiodophenol, Ancylol, DNP, Disofen, Disophenol, Iodofen, Iodophen, Syngamix
InChI Formula InChI=1S/C6H3I2NO3/c7-4-1-3(9(11)12)2-5(8)6(4)10/h1-2,10H
SMILES O=[N](C1=CC(I)=C(O)C(I)=C1)=O
Molecular Weight (g/mol) 390.9
Formula Weight 390.8202