Läs mer
2,6-Diiodophenol, TRC
Produktkod. 30376973
Change view
Kvantitet:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30376973 10 mg
30381204 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30376973 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD45892010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 28177-54-0
Molekylformel C6 H4 I2 O
Molekylvikt (g/mol) 345.9
Synonym Phenol, 2,6-diiodo-, 2,6-Diiodophenol
IUPAC-namn 2,6-diiodophenol
LEDER Oc1c(I)cccc1I
Specifikationer
Kemiskt namn eller material 2,6-Diiodophenol
Kvantitet 10 mg
InChI formel InChI=1S/C6H4I2O/c7-4-2-1-3-5(8)6(4)9/h1-3,9H
Formel vikt 345.8352