Läs mer
2,6-Diisopropyl-1,4-benzoquinone, TRC
Produktkod. 30009494
Change view
Kvantitet:
25 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30009494 100 mg
30007542 25 mg
30004654 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30009494 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD455245100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1988-11-0
Molekylformel C12 H16 O2
Molekylvikt (g/mol) 192.25
Synonym 2,6-Bis(1-methylethyl)benzene-1,4-dione, Propofol Imp. J (EP)
IUPAC-namn 2,6-di(propan-2-yl)cyclohexa-2,5-diene-1,4-dione
LEDER CC(C)C1=CC(=O)C=C(C(C)C)C1=O
Specifikationer
Kemiskt namn eller material 2,6-Bis(1-methylethyl)benzene-1,4-dione
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C12H16O2/c1-7(2)10-5-9(13)6-11(8(3)4)12(10)14/h5-8H,1-4H3
Formel vikt 192.115
Procent renhet >95%