Läs mer
2,6-Dimethoxybenzoyl chloride, 97%, Thermo Scientific™
Product Code. 11398745
Change view
Click to view available options
Kvantitet:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet unitSize
11398745 5 g 5g
11308755 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11398745 Supplier Thermo Scientific Alfa Aesar Supplier No. L12345.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1989-53-3
Molekylformel C9H9ClO3
Molekylvikt (g/mol) 200.62
MDL-nummer MFCD00000665
InChI-nyckel NDXRPDJVAUCBOH-UHFFFAOYSA-N
Synonym benzoyl chloride, 2,6-dimethoxy, 2,6-dimethoxybenzoic acid chloride, 2,6-dimethoxybenzoylchloride, acmc-1bopm, 2,6dimethoxybenzoyl chloride, 2,6-dimethoxy-benzoylchlorid, 2,6-dimethoxylbenzoyl chloride, 2,6-dimethoxy benzoyl chloride, 2,6-dimethoxy-benzoyl chloride, benzoyl chloride,2,6-dimethoxy
PubChem CID 74810
IUPAC-namn 2,6-dimethoxybenzoyl chloride
LEDER COC1=CC=CC(OC)=C1C(Cl)=O
Specifications
Kemiskt namn eller material 2,6-Dimethoxybenzoyl chloride
Smältpunkt 63°C to 67°C
Odör Acrid
Kvantitet 5 g
UN-nummer UN3261
Beilstein 881245
Känslighet Moisture sensitive
Formel vikt 200.62
Procent renhet 97%
Product Title
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.