Learn More
2,6-Dimethyl-1,4-dioxane (>85%), TRC
Product Code. 30385325
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. Quantity Unit Size
30385325 100 mg 100mg
1 options
Product Code. 30385325 Supplier Toronto Research Chemicals Supplier No. TRCD479885100MG
 Restricted Item
Beräknad leverans: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2,6-Dimethyl-1,4-dioxane
CAS 10138-17-7
Molecular Formula C6 H12 O2
Quantity 100 mg
Synonym 2,6-Dimethyl-1,4-Dioxane
InChI Formula InChI=1S/C6H12O2/c1-5-3-7-4-6(2)8-5/h5-6H,3-4H2,1-2H3
SMILES CC1COCC(C)O1
IUPAC Name 2,6-dimethyl-1,4-dioxane
Molecular Weight (g/mol) 116.158
Formula Weight 116.084