Läs mer
2,6-Dimethyl-4-piperidinol (Mixture of Diastereomers), TRC
Produktkod. 30377230
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30377230 500 mg
1 options
Produktkod. 30377230 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD465905500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2,6-Dimethyl-4-piperidinol (Mixture of Diastereomers)
CAS 4733-70-4
Molekylformel C7H15NO
Kvantitet 500 mg
Synonym 2,6-Dimethyl-4-hydroxypiperidine, 2,6-Dimethyl-piperidin-4-ol, 2,6-Lupetidin-4-ol, 4-Hydroxy-2,6-dimethylpiperidine
InChI formel InChI=1S/C7H15NO/c1-5-3-7(9)4-6(2)8-5/h5-9H,3-4H2,1-2H3
LEDER N#Cc1c(F)cccc1
Molekylvikt (g/mol) 129.2