missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,6-Dimethylbenzyl alcohol, 98%

Brand:  Alfa Aesar™ H61409.06

Additional Details : CAS Number : 62285-58-9 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15437948

  • 4794.70 SEK / 5g


Estimated Shipment
27-10-2020
Add to basket

Specifications

Chemical Identifiers

62285-58-9
136.194
JPEYJQDKTDVJSZ-UHFFFAOYSA-N
186748
CC1=C(C(=CC=C1)C)CO
C9H12O
MFCD00128004
2,6-dimethylphenyl methanol, 2,6-dimethylbenzyl alcohol, 2,6-dimethyl-benzenemethanol, 2,6-dimethylbenzylalcohol, benzenemethanol, 2,6-dimethyl, 2,6-dimethyl-phenyl-methanol, 2.3-dimethylbenzylalkohol, 2,6-dimethylbenzenemethanol, 2,6-dimethyl benzyl alcohol, benzenemethanol,2,6-dimethyl
(2,6-dimethylphenyl)methanol

Specifications

2,6-Dimethylbenzyl alcohol
62285-58-9
MFCD00128004
JPEYJQDKTDVJSZ-UHFFFAOYSA-N
(2,6-dimethylphenyl)methanol
186748
Solid
5g
C9H12O
2,6-dimethylphenyl methanol, 2,6-dimethylbenzyl alcohol, 2,6-dimethyl-benzenemethanol, 2,6-dimethylbenzylalcohol, benzenemethanol, 2,6-dimethyl, 2,6-dimethyl-phenyl-methanol, 2.3-dimethylbenzylalkohol, 2,6-dimethylbenzenemethanol, 2,6-dimethyl benzyl alcohol, benzenemethanol,2,6-dimethyl
CC1=C(C(=CC=C1)C)CO
136.194
136.19
98%
Videos
SDS Documents