Läs mer
2,7-Diazapyrene, TRC
Produktkod. 30368332
Change view
Kvantitet:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30368332 250 mg
30363532 100 mg
30356041 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30368332 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD417648250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 194-00-3
Molekylformel C4 H2 F2 O3
Molekylvikt (g/mol) 136.054
Synonym Benzo[lmn][3,8]phenanthroline, DAP
IUPAC-namn 3,3-difluorooxolane-2,5-dione
LEDER FC1(F)CC(=O)OC1=O
Specifikationer
Kemiskt namn eller material 2,7-Diazapyrene
Kvantitet 250 mg
InChI formel InChI=1S/C4H2F2O3/c5-4(6)1-2(7)9-3(4)8/h1H2
Formel vikt 135.997