Läs mer
2,7-Dichlorofluorenyl Bromomethyl Ketone, TRC
Produktkod. 30370311
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30370311 50 mg
1 options
Produktkod. 30370311 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD43448050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2,7-Dichlorofluorenyl Bromomethyl Ketone
CAS 53221-22-0
Molekylformel C15H9BrCl2O
Kvantitet 50 mg
Synonym 2-Bromo-1-(2,7-dichloro-9H-fluoren-4-yl)ethanone, Bromomethyl 2,7-Dichloro-4-fluorenyl Ketone
LEDER Clc1ccc2c(Cc3cc(Cl)cc(C(=O)CBr)c23)c1
Molekylvikt (g/mol) 356.04
Formel vikt 353.9214