missing translation for 'onlineSavingsMsg'
Läs mer

2,8-Bis-(trifluoromethyl)-4-quinolinecarboxylic Acid, Mikromol™

Produktkod. 30050210
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30050210 100 mg
30052664 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30050210 Leverantör Mikromol Leverantörsnummer MM1198.02

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2,8-Bis-(trifluoromethyl)-4-quinolinecarboxylic Acid
TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material Carboxymefloquine
CAS 35853-50-0
Analyt- eller komponentnamn Carboxymefloquine
Frakt skick Room Temperature
Molekylformel C12 H5 F6 N O2
Certifikat och efterlevnad ISO 17025
Synonym 4-Quinolinecarboxylic acid, 2,8-bis(trifluoromethyl)-Ro 21-5104
InChI formel InChI=1S/C12H5F6NO2/c13-11(14,15)7-3-1-2-5-6(10(20)21)4-8(12(16,17)18)19-9(5)7/h1-4H,(H,20,21)
LEDER OC(=O)c1cc(nc2c(cccc12)C(F)(F)F)C(F)(F)F
IUPAC-namn 2,8-bis(trifluoromethyl)quinoline-4-carboxylic acid
Molekylvikt (g/mol) 309.16
Kvantitet 100 mg
Formel vikt 309.0224 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.