Läs mer
2-(8-Chloroperfluorooctyloxy)tetrafluoroethanesulfonyl Fluoride (Technical Grade), TRC
Produktkod. 30350020
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30350020 500 mg
1 options
Produktkod. 30350020 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC351160500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-(8-Chloroperfluorooctyloxy)tetrafluoroethanesulfonyl Fluoride (Technical Grade)
CAS 73606-15-2
Molekylformel C10ClF21O3S
Kvantitet 500 mg
Synonym 2-((8-Chloro-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-hexadecafluorooctyl)oxy)-1,1,2,2-tetrafluoroethanesulfonyl Fluoride
InChI formel InChI=1S/C10ClF21O3S/c11-7(24,25)5(20,21)3(16,17)1(12,13)2(14,15)4(18,19)6(22,23)8(26,27)35-9(28,29)10(30,31)36(32,33)34
LEDER FC(F)(Cl)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)OC(F)(F)C(F)(F)S(=O)(=O)F
IUPAC-namn 2-(8-chloro-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-hexadecafluorooctoxy)-1,1,2,2-tetrafluoroethanesulfonyl fluoride
Molekylvikt (g/mol) 634.59