Läs mer
2,8-Dihydroxyadenine, TRC
Produktkod. 30136778
Change view
Kvantitet:
2.5 mg
25 mg
Förpackningsstorlek:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30136778 25 mg -
30005425 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30136778 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD45010525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 30377-37-8
Molekylformel C5 H5 N5 O2
Molekylvikt (g/mol) 167.13
Synonym 6-Amino-7,9-dihydro-2H-purine-2,8(3H)-dione, 6-amino-1H-Purine-2,8(3H,7H)-dione, 6-Amino-purine-2,8-diol, 2,8-Dihydroxyadenine, 2,8-Dioxyadenine, 6-Amino-1H-purine-2,8(3H,7H)-dione, 6-Amino-2,8-dihydroxypurine
IUPAC-namn 6-amino-7H-purine-2,8-diol
LEDER Nc1nc(O)nc2nc(O)[nH]c12
Specifikationer
Kemiskt namn eller material 2,8-Dihydroxyadenine
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C5H5N5O2/c6-2-1-3(9-4(11)7-1)10-5(12)8-2/h(H5,6,7,8,9,10,11,12)
Formel vikt 167.0443
Procent renhet >95%