missing translation for 'onlineSavingsMsg'
Learn More

2,9-Diacetylguanine, 96%, ACROS Organics™

Brand:  Acros Organics 459211000

Additional Details : CAS Number : 3056-33-5 Weight : 0.10000kg


Quantity 100g

 View more versions of this product

Product Code. 15570604

  • 1543.00 SEK / 100g


Estimated Shipment
09-12-2020
Add to basket

Description

Description

Specifications

Chemical Identifiers

3056-33-5
235.2
n2,9-diacetylguanine, n,9-diacetylguanine, n 2 ,9-diacetylguanine, 2,9-diacetylguanine, 2,9-diacetateguanine, n-9-acetyl-6-oxo-6,9-dihydro-1h-purin-2-yl acetamide, unii-vuw2b9yr95, 2,9-diacetyl guanine, vuw2b9yr95, n-9-acetyl-6-oxo-3h-purin-2-yl acetamide
N-(9-acetyl-6-oxo-3H-purin-2-yl)acetamide
C9H9N5O3
GILZZWCROUGLIS-UHFFFAOYSA-N
76461
CC(=O)NC1=NC(=O)C2=C(N1)N(C=N2)C(=O)C

Specifications

2,9-Diacetylguanine
285.0°C
3056-33-5
n2,9-diacetylguanine, n,9-diacetylguanine, n 2 ,9-diacetylguanine, 2,9-diacetylguanine, 2,9-diacetateguanine, n-9-acetyl-6-oxo-6,9-dihydro-1h-purin-2-yl acetamide, unii-vuw2b9yr95, 2,9-diacetyl guanine, vuw2b9yr95, n-9-acetyl-6-oxo-3h-purin-2-yl acetamide
CC(=O)NC1=NC(=O)C2=C(N1)N(C=N2)C(=O)C
235.2
235.2
Authentic
100g
C9H9N5O3
GILZZWCROUGLIS-UHFFFAOYSA-N
N-(9-acetyl-6-oxo-3H-purin-2-yl)acetamide
76461
96%
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2,9-Diacetylguanine

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS
Documents