Läs mer
2-Acetamido-2-deoxy-3-O-methyl-D-glucopyranose, TRC
Produktkod. 30315424
Change view
Kvantitet:
25 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30315424 25 mg
30316678 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30315424 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA15837525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Acetamido-2-deoxy-3-O-methyl-D-glucopyranose
CAS 94825-74-8
Molekylformel C9 H17 N O6
Kvantitet 25 mg
InChI formel InChI=1S/C9H17NO6/c1-4(12)10-6-8(15-2)7(13)5(3-11)16-9(6)14/h5-9,11,13-14H,3H2,1-2H3,(H,10,12)/t5-,6-,7-,8-,9?/m1/s1
LEDER CO[C@H]1[C@H](O)[C@@H](CO)OC(O)[C@@H]1NC(=O)C
IUPAC-namn N-[(3R,4R,5S,6R)-2,5-dihydroxy-6-(hydroxymethyl)-4-methoxyoxan-3-yl]acetamide
Molekylvikt (g/mol) 235.23
Formel vikt 235.1056