Läs mer
2-Acetamido-2-deoxy-D-glucuronic Acid, TRC
Produktkod. 30179395
Change view
Kvantitet:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30179395 25 mg
30179184 50 mg
30178967 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30179395 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA15827025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 34047-66-0
Molekylformel C8 H13 N O7
Molekylvikt (g/mol) 235.191
Synonym 2-(Acetylamino)-2-deoxy-D-glucuronic Acid, 2-Acetamido-2-deoxy-glucuronic Acid, D-2-Acetamido-2-deoxy-glucuronic Acid, N-Acetyl-D-glucosaminuronic Acid, NSC 256961
IUPAC-namn (2S,3S,4R,5R)-5-acetamido-3,4,6-trihydroxyoxane-2-carboxylic acid
LEDER CC(=O)N[C@H]1C(O)O[C@@H]([C@@H](O)[C@@H]1O)C(=O)O
Specifikationer
Kemiskt namn eller material 2-Acetamido-2-deoxy-D-glucuronic Acid
Kvantitet 25 mg
InChI formel InChI=1S/C8H13NO7/c1-2(10)9-3-4(11)5(12)6(7(13)14)16-8(3)15/h3-6,8,11-12,15H,1H3,(H,9,10)(H,13,14)/t3-,4-,5+,6+,8?/m1/s1
Formel vikt 235.069